Assay Details
Gene Symbol: Mdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031151.2 | NP_112413.2 | 6 - 7 | 736 | 76 | |
GenBank mRNA | BC063165.1 | - | 6 - 7 | 736 | 76 | |
DQ402951.1 | - | 5 - 6 | 570 | 76 | ||
FQ213494.1 | - | 6 - 7 | 75 | |||
FQ213817.1 | - | 6 - 7 | 651 | 76 | ||
FQ215385.1 | - | 6 - 7 | 696 | 76 | ||
FQ216782.1 | - | 6 - 7 | 727 | 76 | ||
FQ217232.1 | - | 6 - 7 | 688 | 76 | ||
FQ224847.1 | - | 6 - 7 | 633 | 76 | ||
X04240.1 | - | 6 - 7 | 677 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
