Assay Details
Gene Symbol: Sh3gl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031238.2 | NP_112517.1 | 7 - 8 | 1077 | 79 | |
XM_006229480.2 | XP_006229542.1 | 7 - 8 | 865 | 79 | ||
XM_006229481.2 | XP_006229543.1 | 7 - 8 | 958 | 79 | ||
XM_006229483.2 | XP_006229545.1 | 7 - 8 | 1064 | 79 | ||
XM_006229485.1 | XP_006229547.1 | 5 - 6 | 608 | 79 | ||
XM_006229486.2 | XP_006229548.1 | 5 - 6 | 532 | 79 | ||
GenBank mRNA | AB008159.2 | - | 4 - 5 | 522 | 79 | |
AF009604.1 | - | 5 - 6 | 561 | 79 | ||
AF227439.1 | - | 7 - 8 | 1080 | 79 | ||
BC078800.1 | - | 7 - 8 | 1080 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
