Assay Details
Gene Symbol: Mapk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031622.2 | NP_113810.2 | 2 - 3 | 1318 | 83 | |
XM_006243409.2 | XP_006243471.1 | 2 - 3 | 1812 | 83 | ||
GenBank mRNA | BC109380.1 | - | 2 - 3 | 1269 | 83 | |
M64301.1 | - | 1 - 2 | 946 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
