Assay Details
Gene Symbol: Casp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031632.1 | NP_113820.1 | 2 - 3 | 585 | 74 | |
GenBank mRNA | AF262319.1 | - | 2 - 3 | 585 | 74 | |
AF271996.1 | - | 2 - 3 | 552 | 74 | ||
AF286006.1 | - | 2 - 3 | 585 | 74 | ||
AF308469.1 | - | 2 - 3 | 552 | 74 | ||
AY008275.1 | - | 2 - 3 | 552 | 74 | ||
AY027667.1 | - | 2 - 3 | 561 | 74 | ||
AY124461.1 | - | 2 - 3 | 585 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
