Assay Details
Gene Symbol: Casp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031632.1 | NP_113820.1 | 8 - 9 | 1324 | 89 | |
GenBank mRNA | AF262319.1 | - | 8 - 9 | 1324 | 89 | |
AF271996.1 | - | 8 - 9 | 1291 | 89 | ||
AF286006.1 | - | 8 - 9 | 1324 | 89 | ||
AF308469.1 | - | 8 - 9 | 1291 | 89 | ||
AY027667.1 | - | 8 - 9 | 1300 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
cellular response to DNA damage stimulus
aging
intrinsic apoptotic signaling pathway in response to DNA damage
response to UV
response to organic cyclic compound
response to cobalt ion
response to estradiol
response to lipopolysaccharide
glial cell apoptotic process
cellular response to UV
signal transduction in response to DNA damage
positive regulation of apoptotic process
positive regulation of neuron apoptotic process
response to antibiotic
cellular response to organic cyclic compound
cellular response to dexamethasone stimulus
regulation of response to DNA damage stimulus
cysteine-type endopeptidase activity
protein binding
peptidase activity
cysteine-type peptidase activity
SH3 domain binding
protein kinase binding
proteolysis
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
cellular response to DNA damage stimulus
aging
intrinsic apoptotic signaling pathway in response to DNA damage
response to UV
response to organic cyclic compound
response to cobalt ion
response to estradiol
response to lipopolysaccharide
glial cell apoptotic process
cellular response to UV
signal transduction in response to DNA damage
positive regulation of apoptotic process
positive regulation of neuron apoptotic process
response to antibiotic
cellular response to organic cyclic compound
cellular response to dexamethasone stimulus
regulation of response to DNA damage stimulus
cysteine-type endopeptidase activity
protein binding
peptidase activity
cysteine-type peptidase activity
SH3 domain binding
protein kinase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P205127 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P238608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132337 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P238608 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P205127 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P205127 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P238608 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P238608 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P205127 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P121227 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P238608 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702079263 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11980 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99100 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72245 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702079263 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10873774 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873776 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873780 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873772 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873770 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873783 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873769 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873781 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873779 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873778 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873784 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873771 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873773 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873785 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873777 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873775 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873782 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10873768 | - | - |
Rat Genome 230 2.0 Array | 1370737_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368652_at | - | - |
Rat Genome 230 2.0 Array | 1370737_at | - | - |
Rat Genome 230 2.0 Array | 1388083_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1368652_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388083_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368652_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388083_a_at | - | - |
Affymetrix HT Rat Focus Array | 1388083_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368652_at | - | - |
Affymetrix HT Rat Focus Array | 1370737_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370737_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500013854.rn.1 | TC0500001835.rn.1 | RTA TC |
Rat Expression Set 230 | 1370737_PM_at | - | - |
Rat Expression Set 230 | 1388083_PM_a_at | - | - |
Rat Expression Set 230 | 1368652_at | - | - |
Rat Expression Set 230 | 1370737_at | - | - |
Rat Expression Set 230 | 1388083_a_at | - | - |
Rat Expression Set 230 | 1368652_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10873768 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20849613 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371435 | - | - |
RatRef-12 Expression BeadChip | ILMN_57734 | - | - |
