Assay Details
Gene Symbol: Foxm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031633.3 | NP_113821.2 | 1 - 2 | 38 | 69 | |
XM_006237418.2 | XP_006237480.1 | 1 - 2 | 38 | 69 | ||
XM_006237420.2 | XP_006237482.1 | 1 - 2 | 38 | 69 | ||
XM_006237421.2 | XP_006237483.1 | 1 - 2 | 38 | 69 | ||
GenBank mRNA | U83112.1 | - | 1 - 2 | 38 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_031633.3, U83112.1, XM_006237418.2, XM_006237420.2, XM_006237421.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
