Assay Details
Gene Symbol: Slc28a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031664.1 | NP_113852.1 | 10 - 11 | 1213 | 66 | |
GenBank mRNA | AY029302.1 | - | 10 - 11 | 1194 | 66 | |
U25055.1 | - | 10 - 11 | 1213 | 66 | ||
U66723.1 | - | 10 - 11 | 1184 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
