Assay Details
Gene Symbol: Aqp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031703.1 | NP_113891.1 | 4 - 5 | 568 | 70 | |
GenBank mRNA | BC127490.1 | - | 4 - 5 | 550 | 70 | |
D17695.1 | - | 4 - 5 | 483 | 70 | ||
L35108.1 | - | 4 - 5 | 568 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
