Assay Details
Gene Symbol: Pdzk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031712.1 | NP_113900.1 | 1 - 2 | 74 | 91 | |
GenBank mRNA | AF013145.1 | - | 1 - 2 | 48 | 91 | |
AF116896.1 | - | 1 - 2 | 74 | 91 | ||
BC078788.1 | - | 1 - 2 | 66 | 7109724 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF013145.1, AF116896.1, BC078788.1, NM_031712.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
