Assay Details
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031719.1 | NP_113907.1 | 1 - 2 | 168 | 70 | |
XM_006229792.2 | XP_006229854.1 | 1 - 2 | 231 | 70 | ||
GenBank mRNA | BC060555.1 | - | 1 - 2 | 212 | 6918578 | 70 |
D13985.1 | - | 1 - 2 | 238 | 70 | ||
D17698.1 | - | 1 - 2 | 151 | 70 | ||
L26450.1 | - | 1 - 2 | 168 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
