Assay Details
Gene Symbol: Htra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031721.1 | NP_113909.1 | 5 - 6 | 1045 | 69 | |
GenBank mRNA | AF179370.1 | - | 5 - 6 | 1045 | 69 | |
BC081767.1 | - | 5 - 6 | 1070 | 69 | ||
FQ220294.1 | - | 2 - 3 | 136 | 69 | ||
FQ228889.1 | - | 4 - 5 | 497 | 69 | ||
FQ228942.1 | - | 4 - 5 | 523 | 69 | ||
FQ229948.1 | - | 2 - 3 | 194 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
