Assay Details
Gene Symbol: Kcnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031742.1 | NP_113930.1 | 1 - 2 | 290 | 83 | |
XM_006250491.2 | XP_006250553.1 | 1 - 2 | 307 | 83 | ||
GenBank mRNA | Z34264.1 | - | 1 - 2 | 290 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
