Assay Details
Gene Symbol: Ceacam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033860.1 | NP_001029032.1 | 2 - 3 | 478 | 106 | |
NM_001033861.1 | NP_001029033.1 | 2 - 3 | 478 | 106 | ||
NM_001033862.1 | NP_001029034.1 | 2 - 3 | 478 | 106 | ||
NM_031755.2 | NP_113943.1 | 2 - 3 | 478 | 106 | ||
GenBank mRNA | AJ277104.1 | - | 2 - 3 | 440 | 106 | |
AJ277105.1 | - | 2 - 3 | 440 | 106 | ||
J04963.1 | - | 2 - 3 | 485 | 106 | ||
M92848.1 | - | 2 - 3 | 478 | 106 | ||
X91137.1 | - | 2 - 3 | 440 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
