Assay Details
Gene Symbol: Cpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031766.1 | NP_113954.1 | 2 - 3 | 193 | 74 | |
GenBank mRNA | AF017637.1 | - | 2 - 3 | 193 | 74 | |
AF017638.1 | - | 2 - 3 | 193 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
