Assay Details
Gene Symbol: Pdxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031769.1 | NP_113957.1 | 4 - 5 | 333 | 66 | |
GenBank mRNA | AF020346.1 | - | 4 - 5 | 333 | 66 | |
FQ213832.1 | - | 4 - 5 | 437 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
