Assay Details
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271984.1 | NP_001258913.1 | 2 - 3 | 95 | 62 | |
NM_031775.3 | NP_113963.2 | 3 - 4 | 250 | 62 | ||
GenBank mRNA | AF025670.1 | - | 3 - 4 | 268 | 62 | |
BC078785.1 | - | 3 - 4 | 215 | 62 | ||
DY310578.1 | - | 2 - 3 | 82 | 62 | ||
FM068086.1 | - | 2 - 3 | 95 | 62 |
Gene Symbol: LOC103689977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761476.1 | XP_008759698.1 | 3 - 4 | 277 | 62 | |
XM_008761477.1 | XP_008759699.1 | 2 - 3 | 100 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
