Assay Details
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031781.1 | NP_113969.1 | 4 - 5 | 920 | 64 | |
XM_006241018.2 | XP_006241080.1 | 5 - 6 | 1081 | 64 | ||
XM_006241019.2 | XP_006241081.1 | 5 - 6 | 1054 | 64 | ||
XM_006241020.2 | XP_006241082.1 | 5 - 6 | 1079 | 64 | ||
GenBank mRNA | AF029109.1 | - | 4 - 5 | 920 | 64 | |
BC161842.1 | - | 5 - 6 | 982 | 9102177 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
