Assay Details
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031793.1 | NP_113981.1 | 4 - 5 | 405 | 98 | |
XM_006234324.2 | XP_006234386.1 | 4 - 5 | 781 | 98 | ||
XM_006234325.2 | XP_006234387.1 | 4 - 5 | 602 | 98 | ||
XM_006234326.2 | XP_006234388.1 | 4 - 5 | 725 | 98 | ||
XM_006234327.2 | XP_006234389.1 | 3 - 4 | 677 | 98 | ||
XM_006234328.2 | XP_006234390.1 | 4 - 5 | 672 | 98 | ||
XM_006234330.2 | XP_006234392.1 | 3 - 4 | 498 | 98 | ||
XM_006234331.2 | XP_006234393.1 | 3 - 4 | 624 | 98 | ||
XM_006234333.2 | XP_006234395.1 | 3 - 4 | 638 | 98 | ||
XM_006234334.2 | XP_006234396.1 | 3 - 4 | 459 | 98 | ||
XM_006234335.2 | XP_006234397.1 | 3 - 4 | 585 | 98 | ||
XM_008761930.1 | XP_008760152.1 | 3 - 4 | 701 | 98 | ||
XM_008761931.1 | XP_008760153.1 | 3 - 4 | 662 | 98 | ||
GenBank mRNA | AF043642.1 | - | 4 - 5 | 405 | 98 | |
BC085723.1 | - | 3 - 4 | 451 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
