Assay Details
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031793.1 | NP_113981.1 | 9 - 10 | 742 | 95 | |
XM_006234324.2 | XP_006234386.1 | 9 - 10 | 1118 | 95 | ||
XM_006234325.2 | XP_006234387.1 | 9 - 10 | 939 | 95 | ||
XM_006234326.2 | XP_006234388.1 | 9 - 10 | 1062 | 95 | ||
XM_006234327.2 | XP_006234389.1 | 8 - 9 | 1014 | 95 | ||
XM_006234328.2 | XP_006234390.1 | 9 - 10 | 1009 | 95 | ||
XM_006234330.2 | XP_006234392.1 | 8 - 9 | 835 | 95 | ||
XM_006234331.2 | XP_006234393.1 | 8 - 9 | 961 | 95 | ||
XM_006234333.2 | XP_006234395.1 | 8 - 9 | 975 | 95 | ||
XM_006234334.2 | XP_006234396.1 | 8 - 9 | 796 | 95 | ||
XM_006234335.2 | XP_006234397.1 | 8 - 9 | 922 | 95 | ||
XM_008761930.1 | XP_008760152.1 | 8 - 9 | 1038 | 95 | ||
XM_008761931.1 | XP_008760153.1 | 8 - 9 | 999 | 95 | ||
GenBank mRNA | AF043642.1 | - | 9 - 10 | 742 | 95 | |
BC085723.1 | - | 8 - 9 | 788 | 95 | ||
FQ225838.1 | - | 4 - 5 | 257 | 95 | ||
FQ230740.1 | - | 4 - 5 | 257 | 95 | ||
FQ230786.1 | - | 4 - 5 | 257 | 95 | ||
FQ232477.1 | - | 4 - 5 | 257 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
