Assay Details
Gene Symbol: Slc1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032065.1 | NP_114454.1 | 1 - 2 | 146 | 79 | |
XM_008765182.1 | XP_008763404.1 | 1 - 2 | 320 | 79 | ||
GenBank mRNA | AY286330.1 | - | 1 - 2 | 140 | 79 | |
BC098912.1 | - | 1 - 2 | 170 | 7315936 | 79 | |
U89608.1 | - | 1 - 2 | 146 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY286330.1, BC098912.1, NM_032065.1, U89608.1, XM_008765182.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
