Assay Details
Gene Symbol: Hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032066.1 | NP_114455.1 | 4 - 5 | 405 | 96 | |
XM_006234599.2 | XP_006234661.1 | 4 - 5 | 572 | 96 | ||
GenBank mRNA | BC061543.1 | - | 4 - 5 | 426 | 6888949 | 96 |
FQ213557.1 | - | 4 - 5 | 441 | 96 | ||
FQ214777.1 | - | 4 - 5 | 470 | 96 | ||
U81186.1 | - | 4 - 5 | 405 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
