Assay Details
Gene Symbol: Hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032066.1 | NP_114455.1 | 9 - 10 | 689 | 116 | |
XM_006234599.2 | XP_006234661.1 | 9 - 10 | 856 | 116 | ||
GenBank mRNA | BC061543.1 | - | 9 - 10 | 710 | 116 | |
FQ213557.1 | - | 9 - 10 | 725 | 116 | ||
FQ214777.1 | - | 9 - 10 | 754 | 116 | ||
U81186.1 | - | 9 - 10 | 689 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
