Assay Details
Gene Symbol: Ppm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270619.1 | NP_001257548.1 | 4 - 5 | 1479 | 121 | |
NM_033096.2 | NP_149087.1 | 4 - 5 | 1479 | 121 | ||
GenBank mRNA | AJ271834.1 | - | 4 - 5 | 1472 | 121 | |
AJ271837.1 | - | 4 - 5 | 1472 | 121 | ||
BC061986.1 | - | 4 - 5 | 1434 | 5599216 | 121 | |
CV110469.1 | - | 2 - 3 | 142 | 121 | ||
FM075941.1 | - | 2 - 3 | 219 | 121 | ||
S90449.1 | - | 4 - 5 | 1765 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
