Assay Details
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033236.1 | NP_150239.1 | 1 - 2 | 84 | 75 | |
GenBank mRNA | BC061542.1 | - | 1 - 2 | 126 | 75 | |
D50694.1 | - | 1 - 2 | 84 | 75 | ||
FQ212821.1 | - | 1 - 2 | 114 | 75 | ||
FQ230973.1 | - | 1 - 2 | 143 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
