Assay Details
Gene Symbol: Ccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053425.1 | NP_445877.1 | 7 - 8 | 722 | 64 | |
GenBank mRNA | AF255305.1 | - | 7 - 8 | 722 | 64 | |
BC158586.1 | - | 7 - 8 | 682 | 64 | ||
EU340033.1 | - | 6 - 7 | 584 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
