Assay Details
Gene Symbol: Fgf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053428.1 | NP_445880.1 | 4 - 5 | 570 | 70 | |
XM_006257592.1 | XP_006257654.1 | 4 - 5 | 838 | 70 | ||
XM_008773572.1 | XP_008771794.1 | 5 - 6 | 746 | 70 | ||
XM_008773573.1 | XP_008771795.1 | 5 - 6 | 808 | 70 | ||
GenBank mRNA | AF271786.1 | - | 4 - 5 | 570 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
neuron migration
apoptotic process
negative regulation of microtubule depolymerization
learning
memory
cell proliferation
hippocampus development
cerebral cortex cell migration
establishment of neuroblast polarity
positive regulation of protein kinase activity
microtubule polymerization
negative regulation of collateral sprouting
protein localization to plasma membrane
microtubule binding
growth factor activity
sodium channel regulator activity
protein kinase activator activity
ion channel binding
beta-tubulin binding
MAPK cascade
neuron migration
apoptotic process
negative regulation of microtubule depolymerization
learning
memory
cell proliferation
hippocampus development
cerebral cortex cell migration
establishment of neuroblast polarity
positive regulation of protein kinase activity
microtubule polymerization
negative regulation of collateral sprouting
protein localization to plasma membrane
microtubule binding
growth factor activity
sodium channel regulator activity
protein kinase activator activity
ion channel binding
beta-tubulin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P130713 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P704429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42969 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P704429 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P130713 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P130713 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P704429 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P704429 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P130713 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P704429 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69132 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48876 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94629 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26095 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15362 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79241 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82726 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33345 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3626 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88497 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31673 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29873 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71585 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36094 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23303 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12189 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99622 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10939921 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939926 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939922 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939920 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939923 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939924 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10939919 | - | - |
Rat Genome 230 2.0 Array | 1368114_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368114_at | - | - |
Affymetrix HT Rat Focus Array | 1368114_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368114_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00015719.rn.1 | TC0X00002509.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00015718.rn.1 | TC0X00002509.rn.1 | RTA TC |
Rat Expression Set 230 | 1368114_PM_at | - | - |
Rat Expression Set 230 | 1368114_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939919 | - | - |
Rat Genome U34 Set | rc_AI112097_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21812027 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355999 | - | - |
RatRef-12 Expression BeadChip | ILMN_50240 | - | - |
