Assay Details
Gene Symbol: Ppp1r9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053474.1 | NP_445926.1 | 4 - 5 | 2140 | 88 | |
XM_008768126.1 | XP_008766348.1 | 3 - 4 | 1648 | 88 | ||
GenBank mRNA | AF016252.1 | - | 4 - 5 | 2140 | 88 | |
BC160878.1 | - | 4 - 5 | 1822 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
