Assay Details
Gene Symbol: Dao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053626.1 | NP_446078.1 | 8 - 9 | 905 | 65 | |
GenBank mRNA | AB003400.1 | - | 8 - 9 | 905 | 65 | |
BC088395.1 | - | 7 - 8 | 820 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
