Assay Details
Gene Symbol: Phyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053674.1 | NP_446126.1 | 4 - 5 | 427 | 86 | |
GenBank mRNA | AF121345.1 | - | 4 - 5 | 427 | 86 | |
BC086573.1 | - | 4 - 5 | 422 | 86 | ||
FQ216943.1 | - | 2 - 3 | 259 | 86 | ||
FQ224924.1 | - | 2 - 3 | 259 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
