Assay Details
Gene Symbol: Cntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053879.1 | NP_446331.1 | 18 - 19 | 2510 | 73 | |
GenBank mRNA | U35371.1 | - | 18 - 19 | 2510 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
