Assay Details
Gene Symbol: Cntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053879.1 | NP_446331.1 | 5 - 6 | 598 | 79 | |
XM_008763166.1 | XP_008761388.1 | 1 - 2 | 329 | 79 | ||
GenBank mRNA | U35371.1 | - | 5 - 6 | 598 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
