Assay Details
Gene Symbol: Epb41l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053927.1 | NP_446379.1 | 20 - 21 | 3201 | 70 | |
XM_006245649.2 | XP_006245711.1 | 20 - 21 | 5514 | 70 | ||
XM_006245651.2 | XP_006245713.1 | 19 - 20 | 5016 | 70 | ||
XM_006245652.2 | XP_006245714.1 | 19 - 20 | 4963 | 70 | ||
XM_008767414.1 | XP_008765636.1 | 24 - 25 | 6531 | 70 | ||
XM_008767415.1 | XP_008765637.1 | 17 - 18 | 4856 | 70 | ||
XM_008767416.1 | XP_008765638.1 | 16 - 17 | 4664 | 70 | ||
XM_008767417.1 | XP_008765639.1 | 15 - 16 | 4610 | 70 | ||
GenBank mRNA | AB032827.1 | - | 20 - 21 | 3201 | 70 | |
AB032828.1 | - | 21 - 22 | 3216 | 70 | ||
DQ462202.1 | - | 16 - 17 | 2416 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
