Assay Details
Gene Symbol: Epb41l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053927.1 | NP_446379.1 | 5 - 6 | 619 | 87 | |
XM_006245649.2 | XP_006245711.1 | 5 - 6 | 2986 | 87 | ||
XM_006245651.2 | XP_006245713.1 | 5 - 6 | 2980 | 87 | ||
XM_006245652.2 | XP_006245714.1 | 5 - 6 | 2981 | 87 | ||
XM_008767414.1 | XP_008765636.1 | 5 - 6 | 2992 | 87 | ||
XM_008767415.1 | XP_008765637.1 | 5 - 6 | 2979 | 87 | ||
XM_008767416.1 | XP_008765638.1 | 5 - 6 | 2979 | 87 | ||
XM_008767417.1 | XP_008765639.1 | 5 - 6 | 2979 | 87 | ||
GenBank mRNA | AB032827.1 | - | 5 - 6 | 619 | 87 | |
AB032828.1 | - | 5 - 6 | 1060 | 87 | ||
DQ462202.1 | - | 4 - 5 | 539 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
