Assay Details
Gene Symbol: Ppat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057198.2 | NP_476546.1 | 4 - 5 | 1073 | 69 | |
GenBank mRNA | BC086999.1 | - | 4 - 5 | 1073 | 69 | |
D10853.1 | - | 4 - 5 | 581 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
