Assay Details
Gene Symbol: Cask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022184.1 | NP_071520.1 | 16 - 17 | 1942 | 74 | |
GenBank mRNA | U47110.1 | - | 16 - 17 | 1942 | 74 |
Gene Symbol: LOC100910506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256650.1 | XP_006256712.1 | 3 - 4 | 348 | 74 | |
XM_006256651.1 | XP_006256713.1 | 3 - 4 | 348 | 74 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
