Assay Details
Gene Symbol: Gpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022215.2 | NP_071551.2 | 1 - 2 | 51 | 64 | |
XM_006257376.1 | XP_006257438.1 | 2 - 3 | 253 | 64 | ||
GenBank mRNA | AB002558.1 | - | 1 - 2 | 39 | 64 | |
BC088396.1 | - | 1 - 2 | 51 | 7108237 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
