Assay Details
Gene Symbol: Gpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022215.2 | NP_071551.2 | 5 - 6 | 630 | 76 | |
XM_006257376.1 | XP_006257438.1 | 6 - 7 | 832 | 76 | ||
GenBank mRNA | AB002558.1 | - | 5 - 6 | 618 | 76 | |
BC088396.1 | - | 5 - 6 | 630 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
