Assay Details
Gene Symbol: Aldh9a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022273.2 | NP_071609.2 | 2 - 3 | 325 | 93 | |
GenBank mRNA | AF170918.1 | - | 2 - 3 | 312 | 93 | |
BC074019.1 | - | 2 - 3 | 325 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
