Assay Details
Gene Symbol: Pafah1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022387.3 | NP_071782.1 | 3 - 4 | 302 | 131 | |
XM_006242957.2 | XP_006243019.1 | 3 - 4 | 768 | 131 | ||
XM_006242958.2 | XP_006243020.1 | 3 - 4 | 301 | 131 | ||
XM_008766172.1 | XP_008764394.1 | 3 - 4 | 532 | 131 | ||
GenBank mRNA | AF016048.1 | - | 3 - 4 | 265 | 131 | |
BC081714.1 | - | 3 - 4 | 302 | 131 | ||
FQ212439.1 | - | 3 - 4 | 590 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
