Assay Details
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022403.2 | NP_071798.1 | 8 - 9 | 883 | 81 | |
GenBank mRNA | BC089802.1 | - | 8 - 9 | 879 | 7370033 | 81 |
M55167.1 | - | 8 - 9 | 883 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
