Assay Details
Gene Symbol: Mlst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022404.2 | NP_071799.2 | 5 - 6 | 779 | 60 | |
GenBank mRNA | AF051155.2 | - | 5 - 6 | 778 | 60 | |
BC097319.1 | - | 5 - 6 | 457 | 7455095 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
