Assay Details
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022502.2 | NP_071947.1 | 1 - 2 | 125 | 140 | |
GenBank mRNA | BC126089.1 | - | 1 - 2 | 125 | 140 | |
FQ213474.1 | - | 1 - 2 | 137 | 140 | ||
FQ213911.1 | - | 137 | 140 | |||
L34262.1 | - | 123 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
