Assay Details
Gene Symbol: Cd151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022523.1 | NP_071968.1 | 803 | 65 | ||
XM_006230576.2 | XP_006230638.1 | 946 | 65 | |||
GenBank mRNA | AF192547.2 | - | 803 | 65 | ||
BC072515.1 | - | 6 - 7 | 772 | 65 | ||
FQ222194.1 | - | 823 | 65 |
Gene Symbol: LOC100911730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749017.3 | XP_003749065.1 | 7 - 8 | 863 | 65 | |
XM_006230598.2 | XP_006230660.1 | 7 - 8 | 946 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
