Assay Details
Gene Symbol: Bbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022629.1 | NP_072151.1 | 2 - 3 | 338 | 67 | |
GenBank mRNA | AF160958.1 | - | 2 - 3 | 338 | 67 | |
FQ210746.1 | - | 430 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
