Assay Details
Gene Symbol: Pgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022847.1 | NP_074038.1 | 8 - 9 | 2612 | 105 | |
GenBank mRNA | HM037361.1 | - | 3164 | 105 | ||
HM037362.1 | - | 3164 | 105 | |||
L16922.1 | - | 8 - 9 | 2612 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
ovulation from ovarian follicle
luteinization
epithelial cell maturation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
transcription by RNA polymerase II
positive regulation of cell population proliferation
negative regulation of gene expression
positive regulation of smooth muscle cell migration
diterpenoid metabolic process
intracellular receptor signaling pathway
mammary gland development
response to progesterone
response to cocaine
negative regulation of apoptotic process
steroid hormone mediated signaling pathway
response to estrogen
estrous cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
regulation of epithelial cell proliferation
progesterone receptor signaling pathway
female mating behavior
tertiary branching involved in mammary gland duct morphogenesis
cellular response to gonadotropin stimulus
cellular response to follicle-stimulating hormone stimulus
cellular response to luteinizing hormone stimulus
RNA polymerase II cis-regulatory region sequence-specific DNA binding
DNA-binding transcription factor activity
steroid hormone receptor activity
nuclear receptor activity
signaling receptor binding
steroid binding
protein binding
zinc ion binding
enzyme binding
hormone binding
sequence-specific DNA binding
ovulation from ovarian follicle
luteinization
epithelial cell maturation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
transcription by RNA polymerase II
positive regulation of cell population proliferation
negative regulation of gene expression
positive regulation of smooth muscle cell migration
diterpenoid metabolic process
intracellular receptor signaling pathway
mammary gland development
response to progesterone
response to cocaine
negative regulation of apoptotic process
steroid hormone mediated signaling pathway
response to estrogen
estrous cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
regulation of epithelial cell proliferation
progesterone receptor signaling pathway
female mating behavior
tertiary branching involved in mammary gland duct morphogenesis
cellular response to gonadotropin stimulus
cellular response to follicle-stimulating hormone stimulus
cellular response to luteinizing hormone stimulus
RNA polymerase II cis-regulatory region sequence-specific DNA binding
DNA-binding transcription factor activity
steroid hormone receptor activity
nuclear receptor activity
signaling receptor binding
steroid binding
protein binding
zinc ion binding
enzyme binding
hormone binding
sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P067613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134094 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P067613 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P067613 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P067613 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P205815 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P359587 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68725 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40748 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91496 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 75952 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10907981 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10907982 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10907983 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10907979 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10907978 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10907977 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10907980 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10907985 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10907984 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10907976 | - | - |
Rat Genome 230 2.0 Array | 1387563_at | - | - |
Rat Genome 230 2.0 Array | 1387563_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387563_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387563_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800000393.rn.1 | TC0800000065.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800000395.rn.1 | TC0800000065.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800000394.rn.1 | TC0800000065.rn.1 | RTA TC |
Rat Expression Set 230 | 1387563_at | - | - |
Rat Expression Set 230 | 1387563_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10907976 | - | - |
Rat Genome U34 Set | S69361mRNA_at | - | - |
Rat Genome U34 Set | S64044_s_at | - | - |
Rat Genome U34 Set | S68284cds_at | - | - |
Rat Genome U34 Set | L16922_at | - | - |
Rat Genome U34 Set | rc_AA999082_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21935313 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376977 | - | - |
