Assay Details
Gene Symbol: Ptprq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022925.1 | NP_075214.1 | 1 - 2 | 236 | 138 | |
GenBank mRNA | AF063249.1 | - | 1 - 2 | 236 | 138 |
Gene Symbol: LOC102552564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241319.2 | XP_006241381.1 | 1 - 2 | 240 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF063249.1, NM_022925.1, XM_006241319.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
