Assay Details
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270838.1 | NP_001257767.1 | 5 - 6 | 726 | 89 | |
NM_001270839.1 | NP_001257768.1 | 5 - 6 | 726 | 89 | ||
NM_001270840.1 | NP_001257769.1 | 5 - 6 | 726 | 89 | ||
NM_022941.4 | NP_075230.1 | 5 - 6 | 726 | 89 | ||
XM_006250261.2 | XP_006250323.1 | 5 - 6 | 835 | 89 | ||
XM_008769758.1 | XP_008767980.1 | 5 - 6 | 838 | 89 | ||
XM_008769759.1 | XP_008767981.1 | 5 - 6 | 838 | 89 | ||
GenBank mRNA | AB104736.1 | - | 5 - 6 | 89 | ||
AB105072.1 | - | 5 - 6 | 89 | |||
AB204897.1 | - | 4 - 5 | 89 | |||
AB204898.1 | - | 4 - 5 | 89 | |||
AB204899.1 | - | 4 - 5 | 89 | |||
AB204900.1 | - | 4 - 5 | 89 | |||
AF133094.1 | - | 4 - 5 | 577 | 89 | ||
AF133095.1 | - | 4 - 5 | 89 | |||
BC089763.1 | - | 5 - 6 | 726 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
