Assay Details
Gene Symbol: Aqp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022960.2 | NP_075249.1 | 8 - 9 | 963 | 62 | |
XM_006243362.1 | XP_006243424.1 | 7 - 8 | 1068 | 62 | ||
XM_008766336.1 | XP_008764558.1 | 4 - 5 | 769 | 62 | ||
GenBank mRNA | AF016406.1 | - | 4 - 5 | 742 | 62 | |
BC085731.1 | - | 8 - 9 | 963 | 62 | ||
FQ211101.1 | - | 4 - 5 | 684 | 62 | ||
FQ212527.1 | - | 4 - 5 | 647 | 62 | ||
FQ214432.1 | - | 7 - 8 | 1073 | 62 | ||
FQ219175.1 | - | 4 - 5 | 684 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
