Assay Details
Gene Symbol: Tmeff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023020.2 | NP_075409.1 | 9 - 10 | 1343 | 80 | |
GenBank mRNA | AJ250730.1 | - | 9 - 10 | 1140 | 80 | |
BC129093.1 | - | 9 - 10 | 1343 | 8363015 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P12356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132168 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12356 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12356 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P12356 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12356 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59148 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69841 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52765 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70484 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78830 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6139 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82776 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10868989 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868997 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868996 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868995 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868991 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868987 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868993 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868998 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868994 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868990 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868992 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868988 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10868986 | - | - |
Rat Genome 230 2.0 Array | 1377772_at | - | - |
Rat Genome 230 2.0 Array | 1387850_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377772_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387850_at | - | - |
Affymetrix HT Rat Focus Array | 1387850_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387850_at | - | - |
Affymetrix HT Rat Focus Array | 1377772_at | - | - |
Affymetrix HT Rat Focus Array | 1377772_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500004251.rn.1 | TC0500000624.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500004249.rn.1 | TC0500000624.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500004250.rn.1 | TC0500000624.rn.1 | RTA TC |
Rat Expression Set 230 | 1387850_at | - | - |
Rat Expression Set 230 | 1377772_PM_at | - | - |
Rat Expression Set 230 | 1377772_at | - | - |
Rat Expression Set 230 | 1387850_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868986 | - | - |
Rat Genome U34 Set | rc_AI639427_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21397774 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1348848 | - | - |
RatRef-12 Expression BeadChip | ILMN_53881 | - | - |
