Assay Details
Gene Symbol: Gabre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023091.1 | NP_075579.1 | 1 - 2 | 77 | 104 | |
GenBank mRNA | AF189262.1 | - | 1 - 2 | 77 | 104 | |
AF255612.1 | - | 91 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
